Hand cleanser, Estesol® Lotion PURE
Light duty hand cleanser to remove light industrial soilings, synthetic and petroleum-based oils, greases and lubricants. For use in general industrial and food manufacturing and processing environments. Can be used on sensitive skin.
- Contains glycerine to help prevent skin dryness
- Mild blend of surfactants for fast and effective hand cleaning
- Petroleum solvent-free, silicone-free, perfume-free
: EU Ecolabel
: *The product(s) on this page are outside the scope of the EU Directive 2019/904