Order Entry
Finland
Orders LinkContactUsLinkComponent
BOD bottles
BOD bottles
Supplier:   Hach
BOD bottles
Supplier:  Hach
Certificates
BOD bottles
3 Options Available Below
Product Details & Documents
These are standard bottles, made of borosilicate glass, for BOD analaysis.
  • Flared lip and ground-glass stopper to prevent air entrapment and for water seal
  • Serialised bottles have silk-screened, sequential numbers and include a bar code
  • Frosted marking area
Delivery information: Stoppers are included.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • HACH62100
    BOD bottle, with glass stopper
    BOD bottle, with glass stopper
    300 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 40,70 €
    40,70 €
    1 * 1 Items
       40,70 €
  • HACH62106
    BOD bottles, with glass stopper
    BOD bottles, with glass stopper
    300 ml
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 233,00 €
    233,00 €
    1 * 6 Items
       233,00 €
  • HACH62124
    BOD bottles, with glass stopper
    BOD bottles, with glass stopper
    300 ml
    Log in for availability
    Log in for availability
    1 * 24 Items
    1 * 24 Items - 855,00 €
    855,00 €
    1 * 24 Items
       855,00 €
Snap caps
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Snap over caps, for standard BOD bottles
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 16,80 €
    16,80 €
    1 * 6 Items
       16,80 €