Order Entry
Finland
ContactUsLinkComponent
Ethyl acetate ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Irritant
Health
Ethyl acetate ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9280-R
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Health
Ethyl acetate ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9280-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    400 lb
  • Seq:
    7284

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.9 %
  • Assay (CH₃COOC₂H₅) (by GC, corrected for water):
    ≥99.9 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.001 %
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Titrable Acid (µeq/g):
    ≤0.9
  • Water (by KF, volumetric):
    ≤0.06 %

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
 :GHS08
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171