Order Entry
Finland
ContactUsLinkComponent
NMR tube washer, SP Wilmad-LabGlass
NMR tube washer, SP Wilmad-LabGlass
Catalog # WILMWG-1207-J10
Supplier:  SP Industries
NMR tube washer, SP Wilmad-LabGlass
Catalog # WILMWG-1207-J10
Supplier:  SP Industries
Specifications
  • Description:
    NMR tube washer, fits tube with O-Ø 10 mm and washer connection 24/40 inner joint, for tube length of 177,8 mm
  • Pk:
    1
Specifications
About this item
NMR tube washer is fitted to a filter flask and a inverted sample tube is inserted into the washer, the solvent is introduced into the reservoir using a series of wash bottles. Can perform numerous wash steps and finish by pulling air through the tube.