Saftey ankle boots, lace-up, Rock & Roll, Jaguar UK
Supplier: U-Power
Certificates
About this item
Safety ankle boot made from water resistant leather with PU scuff.
- WingTex air tunnel textile lining material
- AirToe Composite perforated safety toecap with breathable membrane
- Polyethylene footbed with antifungal treatment and good shock absorption
- Soft PU comfort inner layer sole
- Antislip, anti-static and hard wearing PU outer sole