Safety shoes, slip-on, Response Grip
Supplier: U-Power
Certificates
About this item
100% metal-free safety shoes made from breathable and water repellent New Safety Dry material and a slip and abrasion resistant U.Grip68 PUR sole.
- White anatomically shaped Ergodry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- AirToe Composite toecap with breathable membrane (200 J)
Colour: White
Certifications: EN 20345:2011; S2, SRC