Order Entry
Finland
ContactUsLinkComponent
 
illustra™ ExoProStar™ S kits, Cytiva
Catalog # US79050
Supplier:  Cytiva
CAS Number:  
undefined
illustra™ ExoProStar™ S kits, Cytiva
Catalog # US79050
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Enzymatic clean-up
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    illustra™ ExoProStar™ S PCR and Sequence Reaction Clean Up Kit, 500 reactions
  • Description:
    illustra™ ExoProStar™ S PCR and Sequence Reaction Clean Up Kit

Specifications

About this item

The ExoProStar™ S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar™ S improves digestion efficiency with no degradation of the target PCR product. Sample remain completely intact and ready for immediate use in manual or automated processes. For maximum flexibility the enzymes are supplied in two separate tubes.

  • Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes - just two simple pipetting steps are needed to prepare the reaction
  • Fast 15 minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 10 minutes

The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.