Ion exchange papers, grade SG81, Whatman™
WHAT3668-915
- Flow rate:110 mm/30 min
- Description:Sheets
- Seq:01
- Size:460×570 mm
- Thickness:0,27 mm
- Grade:SG81
- Capillary rise:110 mm/30 min
- Pk:25
These ion exchange papers combine cellulose and large pore silica gel for separations in which both partition and adsorption are important, including the separation of phospholipids, steroids, phenols and dyes.
- Simultaneous development of multiple samples on the same sheet
- Sequential development of the same samples with different solvents and/or different concentrations of the same solvent
- Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution
- Used in enzyme assays to separate product from reactant by charge or polarity