Order Entry
Finland
ContactUsLinkComponent
Centrifuge tubes, Falcon®
Centrifuge tubes, Falcon®
Catalog # BDAA352196
Supplier:  Corning
   Low Manufacturing Impact
undefined
Centrifuge tubes, Falcon®
Catalog # BDAA352196
Supplier:  Corning
   Low Manufacturing Impact

Specifications

  • Ø×L:
    17×120 mm
  • Packed:
    50 per bag/500 per case
  • Capacity:
    15 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    500
  • RCF max.:
    12000×g

Specifications

About this item

PP, high clarity, conical, printed graduation, with dome seal PE screw caps, sterile.

  • Dome seal screw cap offer both vented and fully closed positions
  • RNase/DNase-free

Withstands forces up to 12000×g.