Order Entry
Finland
ContactUsLinkComponent
β-Cholestanol for synthesis, Sigma-Aldrich®
β-Cholestanol for synthesis, Sigma-Aldrich®
Catalog # 8.41513.0005
Supplier:  Merck
CAS Number:  
β-Cholestanol for synthesis, Sigma-Aldrich®
Catalog # 8.41513.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    130 - 143 °C
  • Molar Mass:
    388.67 g/mol
  • EC number:
    201-315-8
  • Hill Formula:
    C27H48O
  • HS Code:
    2906 13 10
  • Storage temperature:
    15...25 °C
  • Assay (GC, area%):
    ≥98% (a/a)
  • Melting range (lower value):
    ≥139 °C
  • Melting range (upper value):
    ≤143 °C
  • Spec. rotation α²0/D (c=1 in chloroform):
    22,0 - 25,0 °
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :388.68 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-315-8
CAS Number:80-97-7
Formula:C₂₇H₄₈O
MDL Number:MFCD00066413