Order Entry
Finland
ContactUsLinkComponent
4-Chlorophenol solution 725 mg/ml DIN EN ISO 9562 for AOX determination, Supelco®
4-Chlorophenol solution 725 mg/ml DIN EN ISO 9562 for AOX determination, Supelco®
Catalog # 1.03085.0100
Supplier:  Merck
CAS Number:  
4-Chlorophenol solution 725 mg/ml DIN EN ISO 9562 for AOX determination, Supelco®
Catalog # 1.03085.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Density:
    1.00 g/cm³ (20 °C)
  • pH:
    5 (H₂O, 20 °C)
  • Storage Class:
    10 - 13 Other liquids and solids
  • WGK:
    NWG not water endangering
  • Storage:
    Store at +15°C to +25°C.
  • Purity:
    ≥99.0 %
  • Identity:
    Passes test
  • Halides:
    ≤100 ppm
  • undefined:
    ≤100 ppm

Specification Test Results

About this item

 :
 :128.56 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:106-48-9
Formula:C₆H₅ClO
MDL Number:MFCD00002318