VWR®, Biohazard Specimen Bags
Supplier: VWR Collection
Certificates
About this item
Designed for safe and secure transport of a specimen container and documentation, these 51 µm thick bags feature a pouch to keep the paperwork separate from the container. They have a self-sealing closure and are tamper evident.