Order Entry
Finland
ContactUsLinkComponent
Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®
Environment
Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®
Catalog # 8.18553.0100
Supplier:  Merck
CAS Number:  
Environment
Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®
Catalog # 8.18553.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Solubility:
    0.014 g/l (20 °C)
  • Melting Point:
    -75 °C
  • Molar Mass:
    222.37 g/mol
  • Boiling Point:
    263 - 266 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    96 °C
  • Refractive Index:
    1.4893 (20 °C, 589 nm)
  • Explosion Limit:
    0.7 - 4.5 %(V)
  • Density:
    0.88 g/cm3 (20 °C)
  • Ignition Point:
    230 °C
  • Vapour pressure:
    -1 hPa (20 °C)
  • Identity (IR):
    Passes test
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
 :222.37 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:230-597-5
UN:3082
 :GHS09
Storage Temperature:Ambient
CAS Number:7212-44-4
Formula:C₁₅H₂₆O
MDL Number:MFCD00008911