Anti-SCAF1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6735R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SR-Related CTD Associated Factor 1
- Antigen symbol:SCAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:58506
- Antigen synonyms:SFR19_HUMAN.|SR-related and CTD-associated factor 1|Splicing factor|Serine arginine-rich pre-mRNA splicing factor SR-A1|SR-related-CTD-associated factor|arginine/serine-rich 19|SR-A1|SCAF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
SR-A1 (serine arginine-rich pre-mRNA splicing factor) is a member of the SR protein family, which interacts with the C-terminal domain of the large subunit of RNA polymerase II and involved in pre-mRNA splicing. The SR-A1 gene is located on chromosome 19q13.3-q13.4. Overexpression of SR-A1 is found in aggressive ovarian cancers.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: SCAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat