Culture tubes
Supplier: EP SCIENTIFIC
Certificates
About this item
Culture tubes are designed for the Total Organic Carbon (TOC) testing and sampling.
- USP method 643 testing
- Off-line and grab sampling of high purity water
- Low background is perfect for preparation and storage of standards
- Cleaning validation is simplified and costs are reduced by using the TOC method of testing
All containers have open-top caps, unless otherwise specified, and fluoropolymer resin-lined septa.
Manufactured, tested and packaged by exacting standards, Thermo Scientific™ TOC Certified Containers are certified to contribute <10 ppb or <20 ppb TOC as background.
Vials feature a septum designed to stay inside the cap while sampling and are appropriate for use as autosampler vials or for sampling.
Ordering information: Supplied with Certificate of Analysis.
Specifications
- Catalog No:
- EPSCCT18-TOC
- EPSCCT18-TOC/LL
- EPSC3115-OTWS-2
- EPSCSCT-18100/TOC
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement