Disposable samplers, DispoPipette
Supplier: BURKLE
Certificates
About this item
Disposable sampler for the highest standard in sampling unlike conventional reusable samplers, disposable samplers do not need to be cleaned after use. Simple disposable sampler for taking samples of low viscosity media. Cleaning reusable samplers is a demanding and often complicated task with greasy toxic substances. Sometimes the task is even impossible. Cross contamination is completely eliminated. It has two opening sizes (2,5/5,5 mm) for different viscosities.
- All-layer and target point samples are possible
- Clean room manufactured
- Optionally sterilised by gamma rays