Order Entry
Finland
ContactUsLinkComponent
Anti-MMP8 Rabbit Polyclonal Antibody
Anti-MMP8 Rabbit Polyclonal Antibody
Catalog # BSBTPB9726
Supplier:  Boster Bio
undefined
Anti-MMP8 Rabbit Polyclonal Antibody
Catalog # BSBTPB9726
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 8 (neutrophil collagenase)
  • Antigen symbol:
    MMP8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4317
  • Antigen synonyms:
    HNC|PMNL-CL|CLG1|MMP-8
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Neutrophil collagenase(MMP8) detection. Tested with WB, IHC-P, ELISA in Human.

Type: Primary
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human