Data Logger, Einstein LabMate
About this item
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
- Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
- Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
- Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
Ordering information: Pressure kit must be ordered separately.
2 Options Available Below
Product Details & Documents
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
- Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
- Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
- Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
Ordering information: Pressure kit must be ordered separately.
Specifications for Product Family
Specifications
- Catalog No:
- FOURENLMSEN
- FOURENLM
- Liitäntä:
- USB
- USB
Specifications
Product Family Options
Product Information
- KuvausAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.FOURENLMSENLiitäntäUSB - Specifications:
More
Specifications
Cat. No.FOURENLMLiitäntäUSB
Accessories
Product Information
- Catalog #KuvausAvailabilityPrice
Product Information
- Catalog #KuvausAvailabilityPrice
Product Information
- Catalog #KuvausAvailabilityPrice