Order Entry
Finland
ContactUsLinkComponent
Markers, bel-pen, SP Bel-Art
Markers, bel-pen, SP Bel-Art
Catalog # BELAF133740000
undefined
Markers, bel-pen, SP Bel-Art
Catalog # BELAF133740000
Specifications
  • Colour:
    Black
  • Description:
    Marking pen with felt tip
  • Seq:
    01
  • Pk:
    3
Specifications
About this item
These felt tip marking pens are perfect for labeling all types of glassware and plastics. The permanent and quick drying ink remains waterproof and smudge proof.
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 133 mm long