Anti-RBC Rabbit Polyclonal Antibody
Catalog # ROCK206-4139
Supplier: Rockland Immunochemicals
Specifications
- Antigen symbol:RBC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Guinea pig
- Format:IgG
- Form:Lyophilized
- Storage buffer:Restore with deionized water (or equivalent), 5.0ml
- Concentration:10,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Guinea Pig washed pooled Red Blood Cells (RBC)
- Purification:This product is an IgG fraction antibody purified from polyspecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
- Size:50 mg
- Pk:50 mg
Specifications
About this item
contains 0.01% (w/v) Sodium Azide
Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig