VWR®, Test Sieves, 315×75 mm
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.