About this item
LDPE or PP, translucent with purple screw caps.
Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high-purity applications, as they have high resistance to chemicals (widely used in ophthalmic products like eye droppers and nasal sprays).
Preferred resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impacts (will not break even if it falls down accidentally, whereas PP carboys may lead to cracking at negative temperatures)
LDPE product, will bounce back to their original shape as highly flexible material
Widely used in pharma industries
PP carboys
For sterile water storage, polypropylene (PP) carboys can be autoclaved before refilling to stop recurring bacterial growth.
Excellent chemical resistance
Working temperature 0 to 135 °C
Autoclavable
Related Information
Certificates
Search by Lot NumberAvantor Cell Culture Solutions.pdf VWR Carboys brochure_Update_MR VWR® Lab bric-a-brac
Product Literature
CoC VWR Carboys with Screw Cap, Wide Neck, Round COC_VWRI216-1730_VGP060319 CoC_VWRI216-1730_VGP070618 CoC_VWRI216-1730_VGP080117 COC_VWRI216-1730_VGP181218
spec_VWR Carboy