Order Entry
Finland
ContactUsLinkComponent
Dunnage rack, heavy-duty
Dunnage rack, heavy-duty
Catalog # METOHP52K3
undefined
Dunnage rack, heavy-duty
Catalog # METOHP52K3

Specifications

  • Description:
    Heavy duty dunnage racks
  • Material:
    Metroseal 3
  • W×D×H:
    760×610×368 mm
  • Pk:
    1
  • Weight:
    14 kg

Specifications

About this item

These heavy duty tubular construction is designed to withstand impact and hold heavy loads.

  • Removable top mat lifts off for cleaning
  • Sturdy: posts and shelves mean fast, easy assembly and secure locking
  • Space Saving: platforms to store bulky items off the floor in less space than if regular shelving were used

Construction: Metroseal 3 epoxy with Microban.

Delivery: Supplied with mat, cap and leveling bolt.