Adapter fittings, Cynergy®
Supplier: ENTEGRIS
Certificates
About this item
Cynergy® Clamp-to-Flaretek® adapter. These adapters show standard Entegris® fitting port connections used in conjunction with Cynergy® clamp fittings.
Using reliable Flaretek® fitting technology, these adapters provide a clean, single-fitting transition to other Entegris® critical fluid management products used in ambient temperature utility line and process applications. Maximum temperature and pressure ratings are size dependent for the Flaretek® fitting connection.