Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microbiology Test Systems
Enzymatic Tests
Reagent for oxidase test in swab
Reagent for oxidase test in swab
Reagent for oxidase test in swab
Catalog # SCAR6119025
Supplier:
Scharlau
CAS Number:
Expand image
Reagent for oxidase test in swab
Catalog # SCAR6119025
Supplier:
Scharlau
CAS Number:
Specifications
Pk:
25
Description:
Oxidase test, sterile swabs
Specifications
About this item
Product Details
Sterile
Product Details