Chlamydia swab
Catalog # DELT300299
Supplier: DELTALAB
Specifications
- Sterile:+
- Description:Chlamydia swabs
- Shaft/head:PS/PE
- Cap colour:Yellow
- Pk:600
Specifications
About this item
Transport swab suitable for the detection of Chlamydia.
- Liquid medium
- Sterilised by irradiaton
- Sample is maintained viable for up to 72 hours
Caution: Recommended to first use a dry swab to clear the channel during cervical sampling.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
*The product(s) on this page are outside the scope of the EU Directive 2019/904