Fluoroline® Ultrapure PFA Tubings
Supplier: ENTEGRIS
New Product
Certificates
About this item
Made of PFA, DuPont 440.
- Compliant to SEMI Standard F57-0301, the specification for polymer components used in ultrapure water and liquid chemical distribution systems
- Made of 100% virgin high purity material to provide the lowest extractable ionic and organic levels in PFA materials
- Tubing is produced with specially designed extruders and extrusion processes
- Tight and consistent tolerances, along with lot-to-lot consistency, are maintained using continuous on-line measurements of the FluoroLine® tubing ext. Ø. and wall thickness
Permanent serialised laser marking is a nondestructive and noncontaminating method to identify tube material and ext. Ø. It also allows for 100% traceability to a specific material lot, production date and production inspection record.