ESD safety ankle boots, lace-up, Dimension Pro, Horizon Mid
Supplier: ELTEN
Certificates
About this item
Leather-free anti-slip ESD safety shoes made from hydrophobised microfibre and hydrophobised textile Cordura® (tear and abrasion resistant, dirt and water repellent) upper, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole Dimension Pro.
- Climate regulating, breathable textile lining
- Semi-orthopaedic, anatomically shaped insole supports the natural heel-to-toe movement of the foot
- Replaceable insole; orthopaedic insoles possible
- Soft fleece ESD insole
- Sole prevents twisting and slipping
- Metal-free puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/silver
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.
Specifications
- Catalog No:
- 114-7827
- 114-7825
- 114-7824
- 114-7835
- 114-7834
- 114-7833
- 114-7832
- 114-7831
- 114-7830
- 114-7829
- 114-7828
- 114-7826
- 114-7836
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001