Measuring spoons, stainless steel
Catalog # BURK5334-8000
Supplier: BURKLE
Specifications
- Capacity:1,25 / 2,5 / 5 / 15 ml
- Description:Measuring spoon set
- Pk:1 SET
Specifications
About this item
Stainless steel V2A. Set of four measuring spoons. Ideal for fast and easy measuring from 1,25 to 15 ml.
- High-gloss finished, handle matt polished
- Volume is engraved on the handle of each measuring spoon