Sealing ring for reaction vessels
Catalog # BIBB296-90
Supplier: SciLabware
Specifications
- Description:Sealing ring, silicone/PTFE, for SVL® reaction vessel flanges and lids
- Pk:1
Specifications
About this item
Silicone rubber sheathed with PTFE.
- Guarantee excellent joint seal
- Temperature resistant from −200 to +200 °C
- Excellent chemical resistance
O-rings for flat flanges with groove.