Order Entry
Finland
ContactUsLinkComponent
Anti-Protein painting of fourth Rabbit Polyclonal Antibody
Anti-Protein painting of fourth Rabbit Polyclonal Antibody
Catalog # BSENR-1571-100
Supplier:  Biosensis
undefined
Anti-Protein painting of fourth Rabbit Polyclonal Antibody
Catalog # BSENR-1571-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein painting of fourth
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Epitope:
    Drosophila melanogaster Protein painting of fourth (22-36 aa)
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Zeste-interacting protein 16
  • Immunogen:
    A synthetic peptide from Drosophila melanogaster Protein painting of fourth (22-36 aa) conjugated to KLH.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Protein painting of fourth (Pof) is a probable RNA-binding protein that binds to the fourth chromosome and may bind a RNA that spreads the fourth chromosome (Ref: Swissprot).

Antibody recognizes painting of fourth protein from flies by ELISA. Other applications and other species not yet tested.
Application Information:
ELISA. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Protein painting of fourth
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Drosophila melanogaster Protein painting of fourth (22-36 aa)
Host: Rabbit
Isotype: IgG
Reactivity: