Sleeve protectors, PP/PE, Hopen®
Catalog # HOPE20.400
Supplier: Medicom
Specifications
- Garment application:Laboratory,Food
- Colour:White
- Length:500 mm
- Pk:100
Specifications
About this item
These disposable sleeve protectors made of PP with PE coating protect the arms against dirt.
- Elasticated
- Latex-free
- Suitable for use in protected environments
Ideal for use in non hazardous applications, such as cleaning, maintenance and food processing.
Thickness: 10 µ
Weight: 35 g/m²