Order Entry
Finland
ContactUsLinkComponent
Anti-PDPK1 Rabbit Polyclonal Antibody
Anti-PDPK1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29292
Supplier:  Abnova
Anti-PDPK1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29292
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3-phosphoinositide dependent protein kinase 1
  • Antigen symbol:
    PDPK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5170
  • Antigen synonyms:
    PDK1|PDPK2|PDPK2P|PRO0461
  • Modification:
    phospho-Tyr9
  • Storage buffer:
    PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y9 of human PDPK1.
  • Purification:
    purified by Affinity Chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PDPK1.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100

Type: Primary
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: