Order Entry
Finland
ContactUsLinkComponent
Anti-SCNN1G Rabbit Polyclonal Antibody (ATTO 488)
Anti-SCNN1G Rabbit Polyclonal Antibody (ATTO 488)
  ABNOPAB28804
 :  Abnova
Anti-SCNN1G Rabbit Polyclonal Antibody (ATTO 488)
  ABNOPAB28804
 :  Abnova

 

  • Antigen name:
    Sodium Channel Nonvoltage-gated 1 gamma
  • Antigen symbol:
    SCNN1G
  • Clonality:
    Polyclonal
  • Conjugation:
    ATTO 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    24768
  • Antigen synonyms:
    Epithelial Na(+) channel subunit gamma|ENaCg|SCNEG)|PHA1|Amiloride-sensitive sodium channel subunit gamma (SCNN1G|Gamma-NaCH|SCNEG,|Gamma-ENaC
  • Storage buffer:
    PBS, 50% glycerol and 0,09% sodium azide
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to Scnn1g C-terminus (amino acids 629-650).
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against recombinant Rat Scnn1g.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000

Type:
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: