Anti-SCNN1B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
ABNOPAB28794
: Abnova
- Antigen name:Sodium Channel Nonvoltage-gated 1 Beta
- Antigen symbol:SCNN1B
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:24767
- Antigen synonyms:ENaCb|BESC1|ENaCbeta|SCNEB
- Storage buffer:PBS, 50% glycerol and 0,09% sodium azide
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to Scnn1b C-terminus (amino acids 617-638).
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against recombinant Rat Scnn1b.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Immunoprecipitation Western Blot: 1:1000
Type:
Antigen: SCNN1B
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype:
Reactivity: