Anti-SCNN1A Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
ABNOPAB28782
: Abnova
- Antigen name:Sodium Channel Nonvoltage-gated 1 Alpha
- Antigen symbol:SCNN1A
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25122
- Antigen synonyms:SCNEA|BESC2|ENaCalpha|SCNN1|ENaCa
- Storage buffer:PBS, 50% glycerol and 0,09% sodium azide
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to Scnn1a N-terminus (amino acids 46-68).
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.
Recommended Dilutions: Immunofluorescence: 1:400; Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: SCNN1A
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity: