Anti-CK20 Mouse Monoclonal Antibody [clone: S4]
Catalog # ABNOH00054474-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 20
- Antigen symbol:CK20
- Clonality:Monoclonal
- Clone:S4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:54474
- Antigen synonyms:K20|KRT21|CK-20|CD20|CK20
- Amino acid number:1 to 424
- Sequence:MDFSRRSFHRSLSSSLQAPVVSTVGMQRLGTTPSVYGGAGGRGIRISNSRHTVNYGSDLTGGGDLFVGNEKMAMQNLNDRLASYLEKVRTLEQSNSKLEVQIKQWYETNAPRAGRDYSAYYRQIEELRSQIKDAQLQNARCVLQIDNAKLAAEDFRLKYETERGIRLTVEADLQGLNKVFDDLTLHKTDLEIQIEELNKDLALLKKEHQEEVDGLHKHLGNTVNVEVDAAPGLNLGVIMNEMRQKYEVMAQKNLQEAKEQFERQTAVLQQQVTVNTEELKGTEVQLTELRRTSQSLEIELQSHLSMKESLEHTLEETKARYSSQLANLQSLLSSLEAQLMQIRSNMERQNNEYHILLDIKTRLEQEIATYRRLLEGEDVKTTEYQLSTLEERDIKKTRKIKTVVQEVVDGKVVSSEVKEVEENI
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:KRT20 (AAH31559, 1 a.a. ~ 424 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant KRT20.
Type: Primary
Antigen: CK20
Clonality: Monoclonal
Clone: S4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human