Anti-AVPR1B Rabbit Polyclonal Antibody
ABNOPAB25516
: Abnova
- Antibody type:Primary
- Antigen name:AVP Receptor V1b
- Antigen symbol:AVPR1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Pig,Rabbit,Horse,Gorilla,Human,Bovine,Dog,Rat
- Cross adsorption:No
- Format:Liquid
- Gene ID:553
- Antigen synonyms:VPR3.|Arginine vasopressin receptor 1B|V1BR_HUMAN|Vasopressin V1b receptor|AVPR V3|Vasopressin V3 receptor|AVPR V1b|V1bR|Pituitary vasopressin receptor 3|AVPR3|Arginine vasopressin receptor 3|Avpr1b|Antidiuretic hormone receptor 1b
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 17 amino acids at N-terminal extracellular domain of human AVPR1B.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of AVPR1B.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4 ?g/ml
Type: Primary
Antigen: AVPR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Pig, Rabbit, Rat