Anti-CLOCK Mouse Monoclonal Antibody [clone: 8F7]
Catalog # ABNOMAB10570
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:clock circadian regulator
- Antigen symbol:CLOCK
- Clonality:Monoclonal
- Clone:8F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:9575
- Antigen synonyms:bHLHe8|KAT13D
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CLOCK.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CLOCK.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000
Type: Primary
Antigen: CLOCK
Clonality: Monoclonal
Clone: 8F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human