Anti-TTPA Mouse Monoclonal Antibody [clone: 7B5]
ABNOH00007274-M01
: Abnova
- Antibody type:Primary
- Antigen name:Tocopherol (alpha) transfer protein
- Antigen symbol:TTPA
- Clonality:Monoclonal
- Clone:7B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7274
- Antigen synonyms:alphaTTP|ATTP|TTP1|AVED
- Amino acid number:179 to 278
- Storage buffer:1x PBS, pH 7,4
- Sequence:IAAVLTDSFPLKVRGIHLINEPVIFHAVFSMIKPFLTEKIKERIHMHGNNYKQSLLQHFPDILPLEYGGEEFSMEDICQEWTNFIMKSEDYLSSISESIQ
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:TTPA (NP_000361.1, 179 a.a. ~ 278 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:50 µg
- Pk:50 µg
Mouse monoclonal antibody raised against a partial recombinant TTPA.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: TTPA
Clonality: Monoclonal
Clone: 7B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human