Anti-TPD52L1 Mouse Monoclonal Antibody [clone: d1C5]
Catalog # ABNOMAB1051
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tumor Protein D52-like 1
- Antigen symbol:TPD52L1
- Clonality:Monoclonal
- Clone:d1C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7164
- Antigen synonyms:TPD52L2|MGC8556|TPD52L1|D53|tumor protein D53|hD53|tumor protein D52-like 1|tumor protein D52-like 2
- Amino acid number:1 to 131
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-131 of human TPD52L1.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant TPD52L1.
Recommended Dilutions: Western Blot: 1:500-1:2000
Type: Primary
Antigen: TPD52L1
Clonality: Monoclonal
Clone: d1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human