Anti-NF68 Mouse Monoclonal Antibody [clone: NF-68]
ABNOMAB1507
: Abnova
- Antibody type:Primary
- Antigen symbol:NF68
- Clonality:Monoclonal
- Clone:NF-68
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified pig Neurofilament 68.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native neurofilament 68.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml
Type: Primary
Antigen: NF68
Clonality: Monoclonal
Clone: NF-68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig