Order Entry
Finland
ContactUsLinkComponent
Anti-Dinitrophenol (DNP) Mouse Monoclonal Antibody [clone: B137M]
Anti-Dinitrophenol (DNP) Mouse Monoclonal Antibody [clone: B137M]
  ABNOMAB3978
 :  Abnova
Anti-Dinitrophenol (DNP) Mouse Monoclonal Antibody [clone: B137M]
  ABNOMAB3978
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Dinitrophenol
  • Antigen symbol:
    DNP
  • Clonality:
    Monoclonal
  • Clone:
    B137M
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Dinitrophenol (DNP)
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against Dinitrophenol (DNP).

Recommended Dilutions: Recommend dilutions start at 5 ?g/ml.

Type: Primary
Antigen: Dinitrophenol (DNP)
Clonality: Monoclonal
Clone: B137M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: