Anti-SIT1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SIT-01]
Catalog # ABNOMAB4525
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Signaling threshold regulating transmembrane adaptor 1
- Antigen symbol:SIT1
- Clonality:Monoclonal
- Clone:SIT-01
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:27240
- Antigen synonyms:SIT|SIT1_HUMAN|SHP intracting transmembrane adapter protein|Signaling threshold-regulating transmembrane adapter 1|gp30/40|SHP2-interacting transmembrane adapter protein|Sit1|Suppression-inducing transmembrane adapter 1.|Suppression inducing transmembrane adapter 1
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to partial human SIT1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant SIT1.
Recommended Dilutions: Flow Cytometry: 1:50
Type: Primary
Antigen: SIT1
Clonality: Monoclonal
Clone: SIT-01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human