Anti-NPT Mouse Monoclonal Antibody [clone: 4B4D1]
Catalog # ABNOMAB5540
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sodium/phosphate cotransporter
- Antigen symbol:NPT
- Clonality:Monoclonal
- Clone:4B4D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human NPT.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant NPT.
Recommended Dilutions: Western Blot: 1:500-1:2000; ELISA: 1:10000
Type: Primary
Antigen: NPT
Clonality: Monoclonal
Clone: 4B4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human