Anti-SLC10A2 Goat Polyclonal Antibody
ABNOPAB15665
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 10 (sodium/bile acid cotransporter Family) Member A2
- Antigen symbol:Solute carrier family 10, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:6555
- Antigen synonyms:ASBT, IBAT, ISBT, NTCP2, PBAM
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-YKANGGFQPDEK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human SLC10A2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Goat polyclonal antibody raised aganist synthetic peptide of SLC10A2.
Recommended Dilutions: ELISA: 1:16000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-6 ?g/ml
Type: Primary
Antigen: SLC10A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human