Anti-BAHD1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB15734
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Bromo Adjacent Homology Domain Containing 1
- Antigen symbol:BAHD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:228536
- Antigen synonyms:BAH Domain-Containing Protein 1|KIAA0945|Gm117|AL022997|AW541238
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:AIRKSYQAVERHGETIRVRDTVLLKSGPRKTSTPYVAKISALWENPESGELMMSLLWYYRPEHLQGGRSPSMHEPLQNEVFASRHQDQNSVACIEEKCYVLTFAEYCRFCAMAKRRGEGLPSRKTALVPPSADYSTPPHRTVPEDTDPELVFLCRHVYDFRHGRILKNPQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 170 mouse Bahd1.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant Bahd1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: BAHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse