Order Entry
Finland
Orders LinkContactUsLinkComponent
Anti-API5 Rabbit Polyclonal Antibody

Anti-API5 Rabbit Polyclonal Antibody

100 μg
Catalog # ABNOPAB7951
Supplier:   Abnova
Anti-API5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB7951
Supplier:  Abnova
Anti-API5 Rabbit Polyclonal Antibody
Anti-API5 Rabbit Polyclonal Antibody

100 μg

Price
523,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

Rabbit polyclonal antibody raised against synthetic peptide of API5.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.5-1 ?g/ml

Type: Primary
Antigen: API5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat

Documents

Specifications
  • Size:
    100 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Affinity purification
  • Antibody type:
    Primary
  • Epitope:
    C-Terminal
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 200 ?l of deionized water and concentration will be 500 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Antigen name:
    Apoptosis inhibitor 5
  • Cat. No.:
    ABNOPAB7951
  • Reactivity:
    Rabbit|Human|Rat|Mouse
  • Format:
    Lyophilized
  • Gene ID:
    8539
  • Antigen synonyms:
    AAC-11|AAC11
  • Immunogen:
    A synthetic peptide corresponding to amino acids at C-terminus of human API5.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    API5
  • Storage buffer:
    0,9 mg NaCl, 0,2 mg Na2HPO4 (5 mg BSA, 0,05 mg sodium azide, 0,05 mg Thimerosal)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.