Anti-RAN Goat Polyclonal Antibody
Catalog # ABNOPAB18990
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RAs-related Nuclear protein
- Antigen symbol:RAN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:5901
- Antigen synonyms:OK/SW-cl.81|GTP-binding nuclear protein Ran|RAN|ARA24|GTPase Ran|Androgen receptor-associated protein 24|Ras-like protein TC4
- Storage buffer:0,5 mg/mL Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-YEHDLEVAQTTALP
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at C-terminus of human RAN.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of RAN.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: RAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat