Anti-SLC5A6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26060
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 5 (sodium-dependent vitamin transporter) Member 6
- Antigen symbol:Solute carrier family 5, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rabbit,Horse,Gorilla,Human,Bovine,Dog
- Cross adsorption:No
- Format:Liquid
- Gene ID:8884
- Antigen synonyms:SMVT
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at internal region of human SLC5A6.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLC5A6.
- Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 μg/ml
Type: Primary
Antigen: SLC5A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Rabbit